Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRP2 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518834
Description
This antibody is predicted to react with equine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
CSRP2 is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.
Specifications
CRP2 | |
Polyclonal | |
Unconjugated | |
CSRP2 | |
AW551867; CRP2; CSRP2; cysteine and glycine rich protein 2; cysteine and glycine-rich protein 2; cysteine rich protein 2; cysteine-rich protein 2; Dlp1; DLP-1; Double LIM protein 1; LIM domain only 5, smooth muscle; LIM domain only protein 5; LIM/double zinc-finger protein; LMO5; LMO-5; SmLIM; Smooth muscle cell LIM protein | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
13008, 1466, 29317 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P97314, Q16527, Q62908 | |
CSRP2 | |
Synthetic peptide sequence (ERLGIKPESVQP) corresponding to the internal amino acids of CSRP2 (aa 86-97). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction