Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531036 Supplier Invitrogen™ Supplier No. PA531036
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (88%), Xenopus laevis (87%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CSRP2 is a member of the CSRP family of genes, encoding a group of LIM domain proteins, which may be involved in regulatory processes important for development and cellular differentiation. CRP2 contains two copies of the cysteine-rich amino acid sequence motif (LIM) with putative zinc-binding activity, and may be involved in regulating ordered cell growth. Other genes in the family include CSRP1 and CSRP3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene CSRP2
Gene Accession No. Q16527
Gene Alias AW551867; CRP2; CSRP2; cysteine and glycine rich protein 2; cysteine and glycine-rich protein 2; cysteine rich protein 2; cysteine-rich protein 2; Dlp1; DLP-1; Double LIM protein 1; LIM domain only 5, smooth muscle; LIM domain only protein 5; LIM/double zinc-finger protein; LMO5; LMO-5; SmLIM; Smooth muscle cell LIM protein
Gene Symbols CSRP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human CSRP2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1466
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.