Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRR9 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6797R

Description
CLPTM1L enhances cisplatin-mediated apoptosis, when overexpressed.Specifications
CRR9 | |
Polyclonal | |
Unconjugated | |
CLPTM1L | |
cisplatin resistance related protein CRR9p; Cisplatin resistance-related protein 9; Cleft lip and palate transmembrane protein 1-like protein; CLPTM1 like; CLPTM1L; CLPTM1-like; CLPTM1-like protein; CRR9; CRR9p | |
Rabbit | |
Protein A | |
RUO | |
81037 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96KA5 | |
CLPTM1L | |
KLH conjugated synthetic peptide derived from human CLPTM1L/CRR9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction