Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crry Antibody (TLD-1C11), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311189
Description
Crry Monoclonal specifically detects Crry in Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Functional, Immunoassay.Specifications
Crry | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Functional, Immunoassay | |
Complement component (3b/4b) receptor 1-like, Complement component receptor 1-like protein, Complement receptor type-1 related gene Y, Complement regulatory protein Crry, CR, Cr1l, CRY, MCP, mCR, mCRY, p65, Protein p65 | |
Rat microglial cells | |
50 μg | |
Primary | |
Rat | |
Purified |
Western Blot, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Functional Assay, Immunoassay | |
TLD-1C11 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
12946 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction