Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Crry Monoclonal Antibody (5I2), FITC
GREENER_CHOICE

Catalog No. PIMA517554
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517554 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517554 Supplier Invitrogen™ Supplier No. MA517554
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17554 targets Crry in FACS and IHC (F) applications and shows reactivity with Rat samples. The MA5-17554 immunogen is erythrocytes from a C3 mutated rat.

This gene encodes a membrane-bound inhibitor of complement activation, which contains several tandemly arranged short consensus repeats (SCRs) of approximately 60 amino acids. The encoded protein may play a role in tumor immune surveillance, which is unrelated to complement inhibition. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Crry
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 5I2
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 4-5mg/mL BSA and 0.02% sodium azide
Gene CR1L
Gene Accession No. Q63135
Gene Alias 5I2 antigen; antigen 5I2; complement C3b/C4b receptor 1 like; complement C4b-binding protein CR-1-like protein; complement component (3b/4b) receptor 1-like; complement component 3b/4b receptor 1-like; complement component receptor 1-like protein; Complement receptor related protein; complement receptor type 1; complement regulatory protein Crry; Cr1; CR1L; Crry; Cry; Mcp; mCRY; MGC129536; protein p65
Gene Symbols CR1L
Host Species Mouse
Immunogen Erythrocytes from a C3 mutated rat.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54243
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.