Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Crry Monoclonal Antibody (5I2)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA170032
Description
MA1-70032 detects Crry in rat samples. MA1-70032 has been successfully used in flow cytometry and immunohistochemistry (frozen sections) procedures. The MA1-70032 immunogen is erythrocytes from a C3 mutated rat.
This gene encodes a membrane-bound inhibitor of complement activation, which contains several tandemly arranged short consensus repeats (SCRs) of approximately 60 amino acids. The encoded protein may play a role in tumor immune surveillance, which is unrelated to complement inhibition. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Crry | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q63135 | |
CR1L | |
Erythrocytes from a C3 mutated rat. | |
250 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen) | |
5I2 | |
Unconjugated | |
CR1L | |
5I2 antigen; antigen 5I2; complement C3b/C4b receptor 1 like; complement C4b-binding protein CR-1-like protein; complement component (3b/4b) receptor 1-like; complement component 3b/4b receptor 1-like; complement component receptor 1-like protein; Complement receptor related protein; complement receptor type 1; complement regulatory protein Crry; Cr1; CR1L; Crry; Cry; Mcp; mCRY; MGC129536; protein p65 | |
Mouse | |
Protein G | |
RUO | |
54243 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction