Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Crry Monoclonal Antibody (8A/E6)

Rat Monoclonal Antibody
Supplier: Invitrogen™ MA549035
Description
The monoclonal antibody 8A/E6 recognizes mouse complement receptor type-1 related gene Y (Crry/p65), a type I membrane protein (MW 56-60 kDa).
This gene encodes a membrane-bound inhibitor of complement activation, which contains several tandemly arranged short consensus repeats (SCRs) of approximately 60 amino acids. The encoded protein may play a role in tumor immune surveillance, which is unrelated to complement inhibition. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Crry | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
Q64735 | |
CR1L | |
Mouse Crry fused to mouse IgG2a-Fc. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
Inhibition Assays, Western Blot, Immunocytochemistry | |
8A/E6 | |
Unconjugated | |
CR1L | |
5I2 antigen; antigen 5I2; complement C3b/C4b receptor 1 like; complement C4b-binding protein CR-1-like protein; complement component (3b/4b) receptor 1-like; complement component 3b/4b receptor 1-like; complement component receptor 1-like protein; Complement receptor related protein; complement receptor type 1; complement regulatory protein Crry; Cr1; CR1L; Crry; Cry; Mcp; mCRY; MGC129536; protein p65 | |
Rat | |
Protein G | |
RUO | |
12946 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction