Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Crry Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA545763
Description
Peptide sequence: ALNKWEPELP SCSRVCQPPP DVLHAERTQR DKDNFSPGQE VFYSCEPGYD Sequence homology: Human: 100%.
This gene encodes a membrane-bound inhibitor of complement activation, which contains several tandemly arranged short consensus repeats (SCRs) of approximately 60 amino acids. The encoded protein may play a role in tumor immune surveillance, which is unrelated to complement inhibition. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
Crry | |
Polyclonal | |
Unconjugated | |
CR1L | |
5I2 antigen; antigen 5I2; complement C3b/C4b receptor 1 like; complement C4b-binding protein CR-1-like protein; complement component (3b/4b) receptor 1-like; complement component 3b/4b receptor 1-like; complement component receptor 1-like protein; Complement receptor related protein; complement receptor type 1; complement regulatory protein Crry; Cr1; CR1L; Crry; Cry; Mcp; mCRY; MGC129536; protein p65 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1379 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q2VPA4 | |
CR1L | |
Synthetic peptide directed towards the middle region of human CR1L. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction