Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crry Rabbit anti-Mouse∣Rat, Polyclonal, Bioss
SDP

Catalog No. 501983189
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3189 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3189 Supplier Bioss Supplier No. BS14070R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a membrane-bound inhibitor of complement activation, which contains several tandemly arranged short consensus repeats (SCRs) of approximately 60 amino acids. The encoded protein may play a role in tumor immune surveillance, which is unrelated to complement inhibition. Three alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.

Specifications

Antigen Crry
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CR1L
Gene Accession No. Q63135, Q64735
Gene Alias 5I2 antigen; Antigen 5I2; Complement C4b-binding protein CR-1-like protein; complement component (3b/4b) receptor 1-like; complement component 3b/4b receptor 1-like; Complement component receptor 1-like protein; Complement receptor related protein; complement receptor type 1; complement receptor type 1-like; Complement regulatory protein Crry; Cr1; CR1L; Crry; LOC102157092; MGC129536
Gene Symbols CR1L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Crry.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12946, 54243
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.