Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Crry Antibody (8A/E6), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP311264
Description
Crry Monoclonal specifically detects Crry in Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoassay.Specifications
| Crry | |
| Monoclonal | |
| 0.1 mg/ml | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoassay | |
| Complement component (3b/4b) receptor 1-like, Complement component receptor 1-like protein, Complement receptor type-1 related gene Y, Complement regulatory protein Crry, CR, Cr1l, CRY, MCP, mCR, mCRY, p65, Protein p65 | |
| Mouse Crry fused to mouse IgG2a-Fc | |
| 50 μg | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot, Immunofluorescence, Immunoassay | |
| 8A/E6 | |
| Unconjugated | |
| 0.2 um filtered solution in PBS, 0.1% BSA | |
| Rat | |
| Protein G purified | |
| RUO | |
| 12946 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction