Learn More
CRSP1/TRAP220 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Turkey, Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Pig, Panda, Orangutan,
Specifications
Specifications
| Antigen | CRSP1/TRAP220 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | MED1 |
| Gene Accession No. | Q15648, Q925J9 |
| Gene Alias | ARC205, Activator-recruited cofactor 205 kDa component, DRIP205, PBP, PPAR-binding protein, PPRB, Peroxisome proliferator-activated receptor binding protein, TRIP2, Thyroid hormone receptor associated protein 220, Thyroid hormone receptor-associated prote |
| Gene Symbols | Med1 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.