Learn More
CRSP1/TRAP220 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300793A
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.Specifications
| CRSP1/TRAP220 | |
| Polyclonal | |
| Unconjugated | |
| MED1 | |
| ARC205, Activator-recruited cofactor 205 kDa component, DRIP205, PBP, PPAR-binding protein, PPRB, Peroxisome proliferator-activated receptor binding protein, TRIP2, Thyroid hormone receptor associated protein 220, Thyroid hormone receptor-associated prote | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 19014, 5469 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q15648, Q925J9 | |
| MED1 | |
| Between 1525 and C-term | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.