Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRSP130 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584839
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584839 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584839 Supplier Invitrogen™ Supplier No. PA584839
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PATLRFPLKG LLPYDKDLFE PQTALLRYVL EQPYSRDMVC NMLGLNKQHK QRCPVLEDQL VDLVVYAMER SETEEKFDDG Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

CRSP is ubiquitously expressed in various tissues and functions as a multimeric complex that consists of nine distinct subunits. Several members of the CRSP family share sequence similarity with multiple components of the yeast transcriptional mediator proteins, including CRSP150, which is related to yeast Rgr1, and CRSP70, which is similar to the elongation factor TFIIS. CRSP77 and CRSP150 are also related to proteins within the putative murine mediator complex, while CRSP130 and CRSP34 are largely unrelated to either murine or yeast proteins. CRSP subunits also associate with larger multimeric co-activator complexes, including ARC/DRI, which binds directly to SREBP and nuclear hormone receptors to facilitate transcription, and with NAT, a polymerase II-interacting complex that represses activated transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRSP130
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MED23
Gene Accession No. Q9ULK4
Gene Alias 130 kDa transcriptional co-activator; 130kDa; 133 kDa transcriptional co-activator; 3000002A17Rik; activator-recruited cofactor 130 kDa component; ARC130; Cofactor required for Sp1 transcriptional activation subunit 3; cofactor required for Sp1 transcriptional activation, subunit 3 (130kD); CRSP complex subunit 3; CRSP130; CRSP133; Crsp3; DRIP130; ESTM7; hSur-2; KIAA1216; Med23; Mediator complex subunit 23; mediator complex subunit MED23 variant MED23_i6; mediator complex subunit MED23 variant MED23_i7; mediator of RNA polymerase II transcription subunit 23; mKIAA1216; MRT18; mSur-2; Protein sur-2 homolog; SUR2; SUR-2; Transcriptional coactivator CRSP130; vitamin D3 receptor interacting protein; Vitamin D3 receptor-interacting protein complex 130 kDa component; X83317
Gene Symbols MED23
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MED23. Recombinant protein control fragment (Product #RP-105501).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9439
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.