Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRSP9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590923
Description
Positive Samples: 293T, NIH/3T3, Mouse testis; Cellular Location: Nucleus.
The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. Two transcript variants encoding the same protein have been found for this gene.
Specifications
CRSP9 | |
Polyclonal | |
Unconjugated | |
MED7 | |
1110063B05Rik; Activator-recruited cofactor 34 kDa component; ARC34; Cofactor required for Sp1 transcriptional activation subunit 9; cofactor required for Sp1 transcriptional activation, subunit 9; cofactor required for Sp1 transcriptional activation, subunit 9 (33kD); cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa; CRSP complex subunit 9; Crsp33; crsp9; hMED7; MED7; Mediator complex subunit 7; mediator of RNA polymerase II transcriptio subunit 7-like protein; mediator of RNA polymerase II transcription subunit 7; mediator of RNA polymerase II transcription subunit 7-like; mediator of RNA polymerase II transcription subunit 7-like protein; MGC12284; RNA polymerase transcriptional regulation mediator subunit 7 homolog; Transcriptional coactivator CRSP33; Unknown (protein for MGC:127209); wu:fd99d12; zgc:56241 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
66213, 9443 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.64 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O43513, Q9CZB6 | |
MED7 | |
Recombinant protein (or fragment). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction