Learn More
Invitrogen™ CRSP9 Polyclonal Antibody

Description
Immunogen sequence: KREEKLEDLK LLFVHVHHLI NEYRPHQARE TLRVMMEVQK RQRLETAERF QKHLERVIEM I Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
Specifications
| Antigen | CRSP9 |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MED7 |
| Gene Accession No. | O43513 |
| Gene Alias | 1110063B05Rik; Activator-recruited cofactor 34 kDa component; ARC34; Cofactor required for Sp1 transcriptional activation subunit 9; cofactor required for Sp1 transcriptional activation, subunit 9; cofactor required for Sp1 transcriptional activation, subunit 9 (33kD); cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa; CRSP complex subunit 9; Crsp33; crsp9; hMED7; MED7; Mediator complex subunit 7; mediator of RNA polymerase II transcriptio subunit 7-like protein; mediator of RNA polymerase II transcription subunit 7; mediator of RNA polymerase II transcription subunit 7-like; mediator of RNA polymerase II transcription subunit 7-like protein; MGC12284; RNA polymerase transcriptional regulation mediator subunit 7 homolog; Transcriptional coactivator CRSP33; Unknown (protein for MGC:127209); wu:fd99d12; zgc:56241 |
| Gene Symbols | MED7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.