Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRSP9 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981358
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1358 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1358 Supplier Bioss Supplier No. BS11427R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The activation of gene transcription is a multistep process that is triggered by factors that recognize transcriptional enhancer sites in DNA. These factors work with co-activators to direct transcriptional initiation by the RNA polymerase II apparatus. The protein encoded by this gene is a subunit of the CRSP (cofactor required for SP1 activation) complex, which, along with TFIID, is required for efficient activation by SP1. This protein is also a component of other multisubunit complexes e.g. thyroid hormone receptor-(TR-) associated proteins which interact with TR and facilitate TR function on DNA templates in conjunction with initiation factors and cofactors. Two transcript variants encoding the same protein have been found for this gene.

Specifications

Antigen CRSP9
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MED7
Gene Accession No. O43513
Gene Alias 1110063B05Rik; Activator-recruited cofactor 34 kDa component; ARC34; BOS_8385; Cofactor required for Sp1 transcriptional activation subunit 9; cofactor required for Sp1 transcriptional activation, subunit 9; cofactor required for Sp1 transcriptional activation, subunit 9 (33kD); cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa; CRSP complex subunit 9; Crsp33; crsp9; hMED7; MED7; Mediator complex subunit 7; Mediator of RNA polymerase II transcription subunit 7; mediator of RNA polymerase II transcription subunit 7-like; MGC12284; RNA polymerase transcriptional regulation mediator subunit 7 homolog; Transcriptional coactivator CRSP33; wu:fd99d12; zgc:56241
Gene Symbols MED7
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CRSP9 .
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9443
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.