Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CRSP9 Recombinant Protein

Catalog No. p-3718285 Shop All Abnova Corporation Products
Click to view available options
Quantity:
10 μg
25 μg

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH05250
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 9443
Molecular Weight (g/mol) 51.37
Name CRSP9 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MGEPQQVSALPPPPMQYIKEYTDENIQEGLAPKPPPPIKDSYMMFGNQFQCDDLIIRPLESQGIERLHPMQFDHKKELRKLNMSILINFLDLLDILIRSPGSIKREEKLEDLKLLFVHVHHLINEYRPHQARETLRVMMEVQKRQRLETAERFQKHLERVIEMIQNCLASLPDDLPHSEAGMRVKTEPMDADDSNNCTGQNEHQRENSGHRRDQIIEKDAALCVLIDEMNERP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CRSP33/CRSP9/MGC12284
Common Name MED7
Gene Symbol MED7
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.