Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRTAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595791
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595791 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595791 Supplier Invitrogen™ Supplier No. PA595791
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NWNGPHRLYL QMSTHGKVRF RDIASPKFSM PSPVRTVITA DFDNDQELEI FFNNIAYRSS SANRLFRVIR REHGDPLIEE LNPGDALEPE GRGTGGVVTD FDGDGMLDLI LSHGESMAQP LSVFRGNQGF NNNWLRVVPR TRFGAFARGA KVVLYTKKSG AHLRIIDGGS GYLCEMEPVA HFGLGKDEAS SVEVTWPDGK MVSRNVASGE MNSVLEILYP RDEDTLQDPA PLECGQGFSQ QENGHCMDTN ECIQFPFVCP RDKPVCVNTY GSYRCRTNKK CSRGYEPNED GTACVGTLGQ SPGPRPTTPT AAAATAAAAA AAGAATAAPV LVDGDLNLGS VVKESCEPSC; Positive Samples: Mouse brain; Cellular Location: extracellular matrix, extracellular space.

CRTAC1 encodes a glycosylated extracellular matrix protein that is found in the interterritorial matrix of articular deep zone cartilage. This protein is used as a marker to distinguish chondrocytes from osteoblasts and mesenchymal stem cells in culture. The presence of FG-GAP motifs and an RGD integrin-binding motif suggests that this protein may be involved in cell-cell or cell-matrix interactions. Copy number alterations in this gene have been observed in neurofibromatosis type 1-associated glomus tumors. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRTAC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CRTAC1
Gene Accession No. Q8R555
Gene Alias 2810454P21Rik; 68 kDa chondrocyte-expressed protein; acidic secreted protein in cartilage; ASPIC; ASPIC1; AW047536; cartilage acidic protein 1; Cep68; CEP-68; chondrocyte expressed protein 68 kDa CEP-68; Crtac1; Crtac1B; FLJ10320; Lotus; Protein CRTAC1-B; W307
Gene Symbols CRTAC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 312-661 of human CRTAC1 (NP_0605283).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72832
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.