Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTAC1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982495
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2495 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2495 Supplier Bioss Supplier No. BS12948R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRTAC1 encodes a glycosylated extracellular matrix protein that is found in the interterritorial matrix of articular deep zone cartilage. This protein is used as a marker to distinguish chondrocytes from osteoblasts and mesenchymal stem cells in culture. The presence of FG-GAP motifs and an RGD integrin-binding motif suggests that this protein may be involved in cell-cell or cell-matrix interactions. Copy number alterations in this gene have been observed in neurofibromatosis type 1-associated glomus tumors. Alternative splicing results in multiple transcript variants.

Specifications

Antigen CRTAC1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CRTAC1
Gene Accession No. Q9NQ79
Gene Alias 2810454P21Rik; 68 kDa chondrocyte-expressed protein; acidic secreted protein in cartilage; ASPIC; ASPIC1; AW047536; Cartilage acidic protein 1; CEP68; CEP-68; chondrocyte expressed protein 68 kDa CEP-68; CRTAC1; Crtac1B; FLJ10320; Lotus; Protein CRTAC1-B; W307
Gene Symbols CRTAC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CRTAC1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55118
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.