Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRTAM Antibody (210213), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB16951K
This item is not returnable.
View return policy
Description
CRTAM Monoclonal antibody specifically detects CRTAM in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| CRTAM | |
| Monoclonal | |
| DyLight 488 | |
| CD355 antigen, class I MHC restricted T cell associated molecule, class-I MHC-restricted T cell associated molecule, Class-I MHC-restricted T-cell-associated molecule, cytotoxic and regulatory T cell molecule, cytotoxic and regulatory T-cell molecule | |
| Mouse myeloma cell line NS0-derived recombinant human CRTAM, Ser18-Ser286, Accession # O95727 | |
| 0.5 mL | |
| Immunology | |
| 56253 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry | |
| 210213 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction