Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTAP Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP132884

Catalog No. NBP132884


Only null left
Add to Cart

Description

Description

CRTAP Polyclonal specifically detects CRTAP in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CRTAP
Polyclonal
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
cartilage associated protein, cartilage-associated protein, CASPLEPREL3, leprecan-like 3
Rabbit
47 kDa
0.1 mL
Signal Transduction
10491
Human
IgG
Western Blot, Immunocytochemistry, Immunofluorescence
0.79mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
O75718
CRTAP
Recombinant protein encompassing a sequence within the center region of human CRTAP. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (94%), Rat (91%). Not yet tested in any other species.
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only