Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRTAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542913 Supplier Invitrogen™ Supplier No. PA542913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FDQNDKVMQQ NLVYYQYHRD TWGLSDEHFQ PRPEAVQFFN VTTLQKELYD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 90%.

The protein encoded by this gene is similar to the chicken and mouse CRTAP genes. The encoded protein is a scaffolding protein that may influence the activity of at least one member of the cytohesin/ARNO family in response to specific cellular stimuli. Defects in this gene are associated with osteogenesis imperfecta, a connective tissue disorder characterized by bone fragility and low bone mass.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRTAP
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CRTAP
Gene Accession No. O75718
Gene Alias 5730529N23Rik; cartilage associated protein; cartilage-associated protein; cartilage-associated-like protein; CASP; CRTAP; leprecan-like 3; LEPREL3; OI7; P3h5; prolyl 3-hydroxylase family member 5 (non-enzymatic); RGD1565180; wu:fb47h01; zgc:85621
Gene Symbols CRTAP
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human CRTAP (aa 324-373).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10491
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.