Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRTAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592216 Supplier Invitrogen™ Supplier No. PA592216
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALRAGRAQYE RYSFRSFPRD ELMPLESAYR HALDKYSGEH WAESVGYLEI SLRLHRLLRD SEAFCHRNCS AAPQPEPAAG LASYPELRLF GGLLRRAHCL KRCKQGLPAF RQSQPSREVL ADFQRREPYK FLQFAYFKAN NLPKAIAAAH TFLLKHPDDE MMKRNMAYYK SLPGAEDYIK DLETKSYESL FIRAVRAYNG ENWRTSITDM ELALPDFFKA F; Positive Samples: SW480; Cellular Location: extracellular matrix, extracellular space.

The protein encoded by this gene is similar to the chicken and mouse CRTAP genes. The encoded protein is a scaffolding protein that may influence the activity of at least one member of the cytohesin/ARNO family in response to specific cellular stimuli. Defects in this gene are associated with osteogenesis imperfecta, a connective tissue disorder characterized by bone fragility and low bone mass.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRTAP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CRTAP
Gene Accession No. O75718, Q9CYD3
Gene Alias 5730529N23Rik; cartilage associated protein; cartilage-associated protein; cartilage-associated-like protein; CASP; CRTAP; leprecan-like 3; LEPREL3; OI7; P3h5; prolyl 3-hydroxylase family member 5 (non-enzymatic); RGD1565180; wu:fb47h01; zgc:85621
Gene Symbols CRTAP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 20-240 of human CRTAP (NP_0063621).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10491, 363158, 56693
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.