Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTC-3 Polyclonal Antibody, Abbomax

Catalog No. 502204583
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4583 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4583 Supplier Abbomax Inc Supplier No. 630410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Tonsil Cellular location: Nucleus and cytoplasm.

CRTC-3 Polyclonal antibody specifically detects CRTC-3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen CRTC-3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Crtc3
Gene Accession No. Q6UUV7, Q91X84
Gene Alias 2610312F20Rik; 6332415K15Rik; AI429792; BC011210; CREB regulated transcription coactivator 3; CREB-regulated transcription coactivator 3; Crtc3; RGD1309666; TORC3; TORC-3; transducer of CREB protein 3; transducer of regulated cAMP response element-binding protein (CREB) 3; transducer of regulated cAMP response element-binding protein 3; transducer of regulated CREB protein 3
Host Species Rabbit
Immunogen A synthetic peptide SQP LDE SWP RQQ PPW KDE KHP G derived from human CRTC-3 protein
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 365297, 64784, 70461
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.