Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRTH-2/GPR44 Antibody (301108) [DyLight 650], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB3338W
Description
CRTH-2/GPR44 Monoclonal antibody specifically detects CRTH-2/GPR44 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
CRTH-2/GPR44 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
301108 | |
Flow Cytometry, CyTOF-ready | |
CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 | |
NS0 mouse myeloma cell line transfected with human CRTH-2, Met1-Ser395, Accession # Q9Y5Y4 | |
0.1 mL | |
GPCR, Innate Immunity | |
11251 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction