Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRTH-2/GPR44 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP176755
Description
CRTH-2/GPR44 Polyclonal specifically detects CRTH-2/GPR44 in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CRTH-2/GPR44 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 | |
Rabbit | |
60 kDa | |
0.1 mg | |
GPCR, Innate Immunity | |
11251 | |
Human, Mouse, Rat, Guinea Pig | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
Q9Y5Y4 | |
PTGDR2 | |
Antibody was raised against a 18 amino acid synthetic peptide from near the amino terminus of human CRTH2. The immunogen is located within the first 50 amino acids of CRTH2. Amino Acid Squence: EQMSRLQSHSNTSIRYID | |
Affinity Purified | |
RUO | |
Primary | |
At least two different isoforms of CRTH2 are known to exist; this antibody will detect both isoforms. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction