Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CRTH-2 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB3338T100UG
This item is not returnable.
View return policy
Description
CRTH-2/GPR44 Monoclonal specifically detects CRTH-2/GPR44 in Human samples. It is validated for Flow Cytometry.Specifications
CRTH-2/GPR44 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q9Y5Y4 | |
PTGDR2 | |
NS0 mouse myeloma cell line transfected with human CRTH-2 Met1-Ser395 Accession # Q9Y5Y4 | |
RUO | |
11251 | |
Human | |
IgG2a |
Flow Cytometry | |
301108 | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, GPR44, PGD2R, PTGDR2, putative G-protein coupled receptor 44 | |
Mouse | |
100 μg | |
Primary | |
Detects human CRTH-2. Stains human CRTH-2-transfected cells but not parental cells. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction