Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Crumble Disks™ Animal Diet
SDP

Catalog No. p-4277299
Encompass
Click to view available options
Quantity:
36/Box
72/Box
Flavor:
Banana

Specifications

Product Type Crumble Disks
Calories 116.9
Components Dextrose, Casein, Cellulose, Corn Starch, Sucrose, Mineral Mix, Canola Oil, Maltodextrin, Tablet Binder, Choline Dihydrogen Citrate, Vitamin Mix, Ascorbic Acid, DL-Methionine, BHA, Natural and Artificial Flavors, Acesulfame Potassium, Citric Acid
Flavor Banana
Format Crumbles
For Use With (Application) GLP and TOX studies
Particle Shape Crumble Disks
Quantity 72/Box
Storage Requirements Cool dry place
Diameter (English) 1.75 in.
Diameter (Metric) 1.27 cm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.