Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541255 Supplier Invitrogen™ Supplier No. PA541255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VDLMHQAVPY PSPPLALDPP RRQRQERTVY TESQQKVLEF YFQKDQYPNY Sequence homology: Human: 100%.

The cone-rod homeobox-containing gene (CRX) encodes a transcription factor that coordinates the expression of several photoreceptor genes in the developing retina, including opsin and rhodopsin. Specifically, CRX binds the OTX motif (TAATCC/A) upstream from photoreceptor genes. The CRX gene is also expressed in the pinealocytes of the pineal gland and may regulate pineal circadian activity by controling the expression of melatonin synthesis genes. Furthermore, CRX(-) mice exhibit disruption of circadian rhythms. The human CRX gene maps to chromosome 19q13.3 within the region of the cone-rod dystrophy-2 locus (CORD2). Mutations in the CRX gene are implicated in the visual pathologies of CORD, Leber congenital amaurosis (LCA) and retinitis pigmentosa (RP). All characterized CRX gene mutations produce disease in heterozygotes although there is no known correlation between the pathologic phenotype and genetic mutation. Missense mutations of the CRX gene affect the homeobox domain, whereas frameshift mutations affect the OTX domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRX
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Crx
Gene Accession No. O43186
Gene Alias Cone rod homeobox; cone-rod homeobox; cone-rod homeobox containing; cone-rod homeobox protein; CORD 2; CORD2; CRD; CRX; Crx1; LCA 7; LCA7; orthodenticle homeobox 3; OTX; OTX 3; OTX3
Gene Symbols Crx
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human CRX.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1406
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.