Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRY1 Mouse anti-Human, Unconjugated, Clone: 4H4-1C4, Abnova™

Catalog No. 89335019 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-019 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-019 Supplier Abnova Corporation Supplier No. H00001407M01J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

CRY1 Monoclonal antibody specifically detects CRY1 in Human samples. It is validated for WB-Ce, IHC-P, IF, S-ELISA, ELISA, WB-Re.

Specifications

Antigen CRY1
Applications Immunohistochemistry (Paraffin), Immunofluorescence, ELISA, Western Blot
Classification Monoclonal
Clone 4H4-1C4
Conjugate Unconjugated
Description cryptochrome 1 (photolyase-like)
Gene CRY1
Gene Accession No. BC030519
Gene Alias PHLL1
Host Species Mouse
Immunogen CRY1 (AAH30519, 1 a.a. ∼ 586 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1407
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.