Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRY2 Antibody (OTI1H5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270449FR
Description
CRY2 Monoclonal specifically detects CRY2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
CRY2 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
CRY2 | |
Full length recombinant protein of human CRY2 (NP_0066940) produced in HEK293T cells. | |
0.1 mL | |
1408.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1H5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
cryptochrome 2 (photolyase-like), cryptochrome-2, FLJ10332, growth-inhibiting protein 37, HCRY2, KIAA0658, PHLL2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction