Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRY2 Monoclonal Antibody (1819CT334.92.36.9)
CRY2 Monoclonal Antibody (1819CT334.92.36.9)
Supplier: Thermo Scientific MA537755
Description
CRY2 Monoclonal Antibody for Western Blot

Specifications
CRY2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q49AN0, Q9R194 | |
CRY2 | |
Recombinant protein of human Cry2. | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Western Blot | |
1819CT334.92.36.9 | |
Unconjugated | |
CRY2 | |
AV006279; CRY2; cryptochrome 2 (photolyase-like); cryptochrome circadian clock 2; cryptochrome-2; D130054K12Rik; growth-inhibiting protein 37; HCRY2; Kiaa0658; PHLL2 | |
Mouse | |
Protein G | |
RUO | |
12953, 1408 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction