Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRY2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589135
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589135 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589135 Supplier Invitrogen™ Supplier No. PA589135
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PVGFGRRTDP SGDYIRRYLP KLKAFPSRYI YEPWNAPESI QKAAKCIIGV DYPRPIVNHA ETSRLNIERM KQIYQQLSRY RGLCLLASVP SCVEDLSHPV AEPSSSQAGS MSSAGPRPLP SGPASPKRKL EAAEEPPGEE LSKRARVAEL PTPELPSKDA.

Various biochemical, physiological and behavioral processes display circadian rhythms controlled by an internal biological clock. The central gears driving this clock appear to be composed of an autoregulatory transcription/post translation-based feedback loop. Cryptochrome 1 (CRY1) and 2 (CRY2) are DNA-binding flavoproteins that bear some homology to blue-light receptors and photolyases. In Drosophila, CRY is a photoreceptor for the circadian clock where it binds to the clock component TIM in a light-dependent fashion and blocks its function. Mammalian CRY1 and CRY2 function via light-independent interactions with circadian genes CLOCK and BMAL1, as well as with PER1, PER2, and TIM. They seem to act as light-independent components of the circadian clock and likely regulate Per1 transcriptional cycling via interactions with both the activator and its feedback inhibitors. Mutant mice not expressing the Cry1 or Cry2 protein display accelerated and delayed periodicity of locomotor activity, respectively. It appears that the combination of both proteins working together is essential to synchronize the organism to circadian phases. A critical balance between Cry1 and Cry2 is required for proper clock function; in complete darkness, double-mutant mice present with instantaneous arrhythmicity, indicating the absence of an internal circadian clock.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRY2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CRY2
Gene Accession No. Q49AN0, Q923I8
Gene Alias AV006279; CRY2; cryptochrome 2 (photolyase-like); cryptochrome circadian clock 2; cryptochrome-2; D130054K12Rik; growth-inhibiting protein 37; HCRY2; Kiaa0658; PHLL2
Gene Symbols CRY2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 455-614 of human CRY2 (NP_0669402).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1408, 170917
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.