Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ cry2Ab Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144572
Description
cry2Ab Polyclonal Antibody for Western Blot, ELISA
Promotes colloidosmotic lysis by binding to the midgut epithelial cells of lepidopteran (Manduca sexta) larvae.
Specifications
cry2Ab | |
Polyclonal | |
Unconjugated | |
cryB2; cryIIA | |
Recombinant Bacillus thuringiensis subsp. kurstaki cry2Ab protein (1-633aa). | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
4.06 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction