Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYAA Monoclonal Antibody (OTI2H2), TrueMAB™, OriGene
SDP

Catalog No. 502539638
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-253-9638 30 μL
50-167-5993 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-253-9638 Supplier OriGene Supplier No. TA505529S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRYAA Monoclonal antibody specifically detects CRYAA in Human samples. It is validated for Western Blot

Specifications

Antigen CRYAA
Applications Western Blot
Classification Monoclonal
Clone OTI2H2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DDT
Gene Accession No. P30046
Gene Alias [a]A-crystallin; Acry 1; Acry-1; alpha A crystallin; alpha crystallin A chain; alpha-A-crystallin; alpha-crystallin A chain; Alpha-crystallin A(1-151); Alpha-crystallin A(1-156); Alpha-crystallin A(1-157); Alpha-crystallin A(1-162); Alpha-crystallin A(1-163); Alpha-crystallin A(1-165); Alpha-crystallin A(1-168); Alpha-crystallin A(1-172); Alpha-crystallin A2 chain; Alpha-crystallin A2(1-162); Alpha-crystallin A2(1-168); Alpha-crystallin A2(1-172); CRYA 1; Crya1; Crya-1; CRYAA; CRYAA2; Crystallin Alpha 1; crystallin alpha A; crystallin, alpha 1; crystallin, alpha A; Crystallin, alpha polypeptide A; crystallin, alpha-1; CTRCT9; DAcry-1; Heat shock protein beta-4; HSPB 4; HSPB4; human alphaA-crystallin (CRYA1); lens opacity 18; LOC102724652; lop18; small heat shock protein; wu:fc06h04; zgc:92036
Host Species Mouse
Immunogen Full length human recombinant protein of human DDT produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1652
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.