Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYAB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586313
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586313 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586313 Supplier Thermo Scientific Supplier No. PA586313
Add to Cart
Add to Cart

CRYAB Polyclonal Antibody

CRYAB Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYAB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Cryab
Gene Accession No. P02511, P23927, P23928
Gene Alias 2310001E10Rik; 3526402H21Rik; AACRYA; active transcription factor CREB; alpha B-crystallin; alpha crystallin B; alpha(B)-crystallin; alpha-B-crystallin; Alpha-crystallin B chain; alpha-crystallin B chain-like protein; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CMD1II; CREB; CREB1; CREB-1; Crya2; Crya-2; CRYAB; CryAB antibody; crystallin alpha B; crystallin, alpha 2; crystallin, alpha B; crystallin, alpha polypeptide 2; CTPP2; CTRCT16; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; epididymis secretory protein Li 101; heat shock protein beta-5; heat shock protein CryAB; heat shock protein HspB5; heat-shock 20 kD like-protein; HEL-S-101; HSPB5; HspB5 protein; MFM2; P23; Renal carcinoma antigen NY-REN-27; rosenthal fiber component; transactivator protein; Y protein
Gene Symbols Cryab
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 21-70 of Human Crystallin-alphaB.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12955, 1410, 25420
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.