Learn More
Invitrogen™ CRYBA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528954
Description
Recommended positive controls: 293T, A431, H1299, HeLa, Molt-4, Raji, mouse fetal eye. Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (84%), Dog (96%), Rabbit (96%), Chicken (84%), Rhesus Monkey (97%), Bovine (93%), Guinea pig (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group, none in the acidic group). Beta-crystallins form aggregates of different sizes and are able to self-associate to form dimers or to form heterodimers with other beta-crystallins. This gene, a beta acidic group member, encodes two proteins (crystallin, beta A3 and crystallin, beta A1) from a single mRNA, the latter protein is 17 aa shorter than crystallin, beta A3 and is generated by use of an alternate translation initiation site. Deletion of exons 3 and 4 causes the autosomal dominant disease 'zonular cataract with sutural opacities'.
Specifications
CRYBA1 | |
Polyclonal | |
Unconjugated | |
CRYBA1 | |
BA3/A1; BA3A1C; beta crystallin A3 chain transcript CN; beta crystallin A3 chain transcript LAM; beta crystallin A3 chain transcript PS; beta crystallin A3 chain transcript TC; beta-A1-crystallin; beta-A3; beta-A3 crystallin; beta-A3/A1-crystallin; beta-crystallin A1; beta-crystallin A3; Beta-crystallin A3, isoform A1, Delta4 form; Beta-crystallin A3, isoform A1, Delta7 form; Beta-crystallin A3, isoform A1, Delta8 form; Cryb; CRYB1; Cryba1; crystallin beta A1; crystallin beta A3; crystallin beta A3/A1; crystallin, beta A1; crystallin, beta A3; CTRCT10; eye lens structural protein; truncated beta-crystallin A3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12957, 1411 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.26 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P02525, P05813 | |
CRYBA1 | |
Full length human beta A3 Crystallin Recombinant protein. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.