Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRYBA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584545
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584545 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584545 Supplier Invitrogen™ Supplier No. PA584545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYQYVLERDR HSGEFCTYGE LGTQAHTGQL QSIRRVQH Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.

Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of the vertebrate eye, which function to maintain the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also defined as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group but absent in the acidic group). Beta-crystallins form aggregates of different sizes and are able to form homodimers through self-association or heterodimers with other beta-crystallins. This gene is a beta acidic group member. Three alternatively spliced transcript variants encoding identical proteins have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYBA2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cryba2
Gene Accession No. P53672
Gene Alias beta-A2 crystallin; betaA2-crystallin; Beta-crystallin A2; Cryba2; crystallin beta A2; crystallin, beta A2; CTRCT42; E130107M19Rik; eye lens structural protein; lens structural protein
Gene Symbols Cryba2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRYBA2. Recombinant protein control fragment (Product #RP-108468).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1412
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.