Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRYBA4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | CRYBA4 |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:20 - 1:50 |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CRYBA4 Polyclonal specifically detects CRYBA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CRYBA4 | |
Polyclonal | |
Rabbit | |
Human | |
Beta-A4 crystallin, beta-crystallin A4, crystallin, beta A4, crystallin, beta polypeptide A4, eye lens structural protein, MCOPCT4 | |
CRYBA4 | |
IgG | |
Affinity Purified |
Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Unconjugated | |
RUO | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
1413 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MTLQCTKSAGPWKMVVWDEDGFQG | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title