Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRYBA4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597628
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597628 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597628 Supplier Invitrogen™ Supplier No. PA597628
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRYBA4 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Crystallins are the dominant structural components of the vertebrate eye lens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYBA4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CRYBA4
Gene Accession No. P53673, Q9JJV0
Gene Alias beta crystallin A4 chain transcript PS; beta crystallin alpha 4 chain; beta-A4 crystallin; Beta-crystallin A4; Cryba4; crystallin beta A4; crystallin, beta A4; crystallin, beta polypeptide A4; CTA-445C9.1; CTRCT23; CYRBA4; eye lens structural protein; MCOPCT4
Gene Symbols CRYBA4
Host Species Rabbit
Immunogen Recombinant Human Beta-crystallin A4 protein (2-196AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12959, 1413
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.