Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRYBB2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$202.00 - $357.00
Specifications
| Antigen | CRYBB2 |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
FEPA5145324
|
Invitrogen™
PA5145324 |
20 μL |
Each for $202.00
|
|
|||||
FEPA5145323
![]() |
Invitrogen™
PA5145323 |
200 μL |
Each for $357.00
|
|
|||||
Description
Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Beta-crystallins, the most heterogeneous, differ by the presence of the C-terminal extension (present in the basic group, none in the acidic group). Beta-crystallins form aggregates of different sizes and are able to self-associate to form dimers or to form heterodimers with other beta-crystallins. This gene, a beta basic group member, is part of a gene cluster with beta-A4, beta-B1, and beta-B3. A chain-terminating mutation was found to cause type 2 cerulean cataracts.Specifications
| CRYBB2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat | |
| P62696, P62697 | |
| 12961, 25422 | |
| Recombinant Rat CRYbB2 protein, Pro17-Arg191 (Accession #P62697). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Beta crystallin B2; Beta crystallin Bp; beta-B2 crystallin; Beta-crystallin B2; beta-crystallin Bp; CCA 2; CCA2; CRYB B2; CRYB2; Cryb-2; CRYB2A; CRYBB 2; CRYBB2; crystallin beta B2; crystallin, beta B2; CTA-221G9.7; CTRCT3; D22S665; eye lens structural protein; OTTHUMP00000198622; R.norvegicus CRYBB2 gene (crystallin, beta B2) | |
| Crybb2 | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Crybb2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title