Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRYGD Monoclonal Antibody (2H3)
GREENER_CHOICE

Catalog No. PIMA518655
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518655 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518655 Supplier Invitrogen™ Supplier No. MA518655
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: SVRSCRLIPH SGSHRIRLYE REDYRGQMIE FTEDCSCLQD RFRFNEIHSL NVLEGSWVLY ELSNYRGRQY LLMPGDYRRY QDWGATNARV GSLRRVIDFS.

Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Gamma-crystallins are a homogeneous group of highly symmetrical, monomeric proteins typically lacking connecting peptides and terminal extensions. They are differentially regulated after early development. Four gamma-crystallin genes (gamma-A through gamma-D) and three pseudogenes (gamma-E, gamma-F, gamma-G) are tandemly organized in a genomic segment as a gene cluster. Whether due to aging or mutations in specific genes, gamma-crystallins have been involved in cataract formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYGD
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CRYGD
Gene Accession No. P07320
Gene Alias Aey4; Beta4abc; CACA; CCA3; CCP; Cryg-1; Cryg4; Crygd; cry-g-D; crystallin gamma D; crystallin, gamma D; Crystallin, gamma polypeptide 4; CTRCT4; DGcry-1; gamma crystallin 4; Gamma crystallin D; Gamma D crystallin; gamma-crystallin 1; Gamma-crystallin 2-2; gamma-crystallin 4; Gamma-crystallin D; gamma-D-crystallin; Len; lens opacity 12; Lop12; PCC
Gene Symbols CRYGD
Host Species Mouse
Immunogen CRYGD (NP_008822, 75 a.a. ~ 174 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1421
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.