Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRYGS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557155 Supplier Invitrogen™ Supplier No. PA557155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GTWAVYERPN FAGYMYILPQ GEYPEYQRWM GLNDRLSSCR AVHLPSGGQY KIQIFEKGDF SGQMYETTED CPSIMEQFHM REIHSCKVLE GVWIFYELP Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 92%.

Crystallins are separated into two classes: taxon-specific, or enzyme, and ubiquitous. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. Since lens central fiber cells lose their nuclei during development, these crystallins are made and then retained throughout life, making them extremely stable proteins. Mammalian lens crystallins are divided into alpha, beta, and gamma families; beta and gamma crystallins are also considered as a superfamily. Alpha and beta families are further divided into acidic and basic groups. Seven protein regions exist in crystallins: four homologous motifs, a connecting peptide, and N- and C-terminal extensions. Gamma-crystallins are a homogeneous group of highly symmetrical, monomeric proteins typically lacking connecting peptides and terminal extensions. They are differentially regulated after early development. This gene encodes a protein initially considered to be a beta-crystallin but the encoded protein is monomeric and has greater sequence similarity to other gamma-crystallins. This gene encodes the most significant gamma-crystallin in adult eye lens tissue. Whether due to aging or mutations in specific genes, gamma-crystallins have been involved in cataract formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYGS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRYGS
Gene Accession No. P0C5E9, P22914
Gene Alias AI327013; Beta-crystallin S; beta-crystallin S-like protein; CRYG8; CRYGS; crystallin gamma S; crystallin, gamma 8; crystallin, gamma polypeptide 8; crystallin, gamma S; CTRCT20; Gamma-crystallin S; gamma-crystallin S; beta-crystallin S; gamma-S-crystallin; gammaS-crystallin; opacity due to poor secondary fiber cell junction; Opj; recessive nuclear cataract; rncat; unnamed protein product
Gene Symbols CRYGS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRYGS. Recombinant protein control fragment (Product #RP-96492).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1427, 689897
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.