Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRYGS Polyclonal Antibody
CRYGS Polyclonal Antibody
Supplier: Thermo Scientific PA569836
Description
CRYGS Polyclonal Antibody for Western Blot

Specifications
CRYGS | |
Polyclonal | |
Unconjugated | |
CRYGS | |
AI327013; Beta-crystallin S; beta-crystallin S-like protein; CRYG8; CRYGS; crystallin gamma S; crystallin, gamma 8; crystallin, gamma polypeptide 8; crystallin, gamma S; CTRCT20; Gamma-crystallin S; gamma-crystallin S; beta-crystallin S; gamma-S-crystallin; gammaS-crystallin; opacity due to poor secondary fiber cell junction; Opj; recessive nuclear cataract; rncat; unnamed protein product | |
Rabbit | |
Affinity Chromatography | |
RUO | |
689897 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P0C5E9 | |
CRYGS | |
synthetic peptide directed towards the middle region of human CRYGS (aa 121-170). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction