Learn More
CRYM Mouse anti-Human, Clone: OTI1B5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501433

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Crystallins are separated into two classes: taxon-specific and ubiquitous. The former class is also called phylogenetically-restricted crystallins. The latter class constitutes the major proteins of vertebrate eye lens and maintains the transparency and refractive index of the lens. This gene encodes a taxon-specific crystallin protein that binds NADPH and has sequence similarity to bacterial ornithine cyclodeaminases. The encoded protein does not perform a structural role in lens tissue, and instead it binds thyroid hormone for possible regulatory or developmental roles. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
CRYM | |
Monoclonal | |
Unconjugated | |
CRYM | |
DFNA40, THBP | |
Mouse | |
Affinity Chromatography | |
RUO | |
1428 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1B5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q14894 | |
CRYM | |
Full length human recombit protein of human CRYM produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.