Learn More
United Scientific™ Cryo Coder For Star and Round Base Vials
Shop All United Scientific Supplies, Inc Products
Description
- Designed to fit onto the top of our star and round base cryogenic vial closures.
- Provide quick visual identification of specific vials for ease in organizing inventory.
- Available in a range of colors.
- Compatible with all United Scientific cryo vials except P60102 and P60103 skirted base cryo vials.
Specifications
Specifications
| Color | White |
| Vial Type | United Star and Round Base Cryo Vials |
| Product Type | Cryo Coder |