Learn More
Thermo Scientific™ CryoMed™ Sensors
Catalog No. 13262826
Change view
Click to view available options
For Use With (Equipment):
0.020 Sheath Straw Sample
0.062 Sheath Vial Sample
1.2/2.0 Cryo Tubes in CryoMed
4.0/5.0 Cryo Tubes in CryoMed
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-262-826 0.020 Sheath Straw Sample
TF4000385 1.2/2.0 Cryo Tubes in CryoMed
11-679-122 4.0/5.0 Cryo Tubes in CryoMed
13-262-827 0.062 Sheath Vial Sample
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 13-262-826 Supplier Thermo Scientific™ Supplier No. 4000402
Only null left
Add to Cart
Add to Cart
Improve the functionality of Thermo Scientific™ Cryomed™ controlled-rate freezers with these CryoMed Sensors.
Features:
  • Thermocouple sensors for cryo tubes, sheath straw and vial samples
  • Includes male plug
Recommended for:
Controlled-rate freezing applications
TRUSTED_SUSTAINABILITY
Specifications
Description Thermocouple Sensor
Type Thermocouple Sensor
For Use With (Equipment) 0.020 Sheath Straw Sample
Plug Type Male
Holds Cryo Tube, Sheath Straw, Vial