Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cryopreserved Rat Kupffer Cells

Catalog No. rtkccs
Encompass_Preferred
Click to view available options
Quantity:
1 vial

Cryopreserved Rat Kupffer Cells

These cryopreserved purified Kupffer cells are isolated from adult male Sprague-Dawley rats. Each vial contains a minimum of one million cells.

Easy to use cryopreserved Kupffer cells provide a convenient way to produce custom cell ratios for hepatic co-culture.

Each lot of Cryopreserved Rat Kupffer Cells is tested for:

  • Purity
  • Response to activation with LPS
  • Adherence
  • Minimum 1 million cells per vial

    Kupffer Cells for Improved Physiological Modeling

    Growing evidence shows that under both normal and pathological conditions, many hepatocyte functions are regulated by substances released from neighboring non-parenchymal cells (NPC). These cells, particularly Kupffer cells, play an important role in the modulation of xenobiotic metabolism in the liver. Kupffer cells secrete potent mediators of the inflammatory response that controls liver inflammation. These cytokine mediators control hepatocyte metabolic rates through direct interactions with phase I and phase II enzymes. Kupffer cell and hepatocyte co-cultures can self-assemble within 72 hours of treatment with pro-inflammatory cytokines or LPS, and can function by effectively modulating P450 expression, thus giving a more physiologically relevant result. Studies indicate that co-culture of hepatocytes with NPCs would better represent both normal liver physiology as well as disease states.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.,

    Note:
    These products require shipment in LN2 vapor dewars therefore additional fees may apply.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Cell Passage Number Passage 0
    Content And Storage 1 vial of 1 million viable rat (Sprague-Dawley) Kupffer cells.

    Store in a cryostorage dewar or −135°C freezer.
    Cell Type Macrophage Cells
    Culture Type Adherent Cell Culture
    Form Cryopreserved
    Gender Male
    Species Rat
    Tissue Liver
    Donor Attributes Normal
    Donor Source Pooled
    Format Tube
    No. of Cells 1 x 106
    Product Line CellzDirect™
    Product Type Rat Kupffer Cell
    Quantity 1 vial
    Tested For Purity, Response to Activation with LPS, Adherence, Minimum 1 Million Cell per Vial
    Show More Show Less
    WARNING: Cancer - www.P65Warnings.ca.gov
    Product Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.