Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ad Infinitum™ CryoProtectPURE STEM™ DMSO-Free/Serum-Free Cryopreservation Media

Catalog No. p-7224576
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

CryoProtectPURE STEM™ DMSO-Free/Serum-Free Cryopreservation Media is optimized for stem cell applications. Premixed, ready-to-use.

CryoProtectPURE STEM™ DMSO-Free/Serum-Free Cryopreservation Media increases cell viability, produces healthier cells post-thaw with no antigenicity concern or toxic vitrification, produces higher repeatability, higher viability, and increased preservation of expression post thaw.

  • DMSO-Free
  • Serum-Free
  • Pre-mixed, ready-to-use, 0.2 μm filtered
  • Optimized for Stem Cell Applications
  • Shipped at ambient temperature.
  • Store at 2 to 8°C.
  • For manufacturing and/or research use only, not for diagnostic or therapeutic use.
  • Manufactured at a cGMP-compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.

Specifications

Product Type Cryopreservation Media
Quantity 25 mL
Content And Storage 2°C to 8°C

For manufacturing and/or research use only, not for diagnostic or therapeutic use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.