Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ CryoStorage Vials, 3D SafeCode

Catalog No. 05413988 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
0.5 mL
1.0 mL
1.5 mL
2.0 mL
4.0 mL
Sterility:
Non-sterile
Sterile
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
05-413-988 0.5 mL Sterile
05-413-983 0.5 mL Non-sterile
05-413-984 1.0 mL Non-sterile
05-413-985 1.5 mL Non-sterile
05-413-986 2.0 mL Non-sterile
05-413-987 4.0 mL Non-sterile
05-413-989 1.0 mL Sterile
05-413-990 1.5 mL Sterile
05-413-991 2.0 mL Sterile
05-413-992 4.0 mL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 05-413-988 Supplier Eppendorf™ Supplier No. 0030079450
Add to Cart
Add to Cart

Provides a 3-level coding of the SafeCode system (datamatrix code, barcode, and clear writing) to enable safe sample identification. Up to 96 pre-labeled vials are pre-racked and pre-capped (0.5 mL to 4.0 mL) for storage applications.

  • 3-level coding for safe sample identification
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Temperature range: -196 °C (gas phase of LN2) to +121 °C
  • Manufactured from high-quality polypropylene in class VIII cleanroom for highest sample purity
  • Pre-capped and pre-racked for comfortable handling
  • Additional information on vials available via Eppendorf website for documentation purposes
  • Direct import into Eppendorf eLABInventory and eLABJournal software for convenient handling
TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Sterility Sterile
Volume (Metric) 0.5 mL
No. per Pack 96
RNase Free Yes
Dnase Free Yes
Barcode 2D SafeCode
Quantity 960 (10 Racks of 96 Vials)
Product Type Coded Cryobank Vial System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.